missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IL-17A; Flex Set, Bead B5

Determines human reactivity by testing samples with BD CBA Human IL-13 Flex Set

Brand:  BD 560383


Product Code. 15800159

  • 409.00€ / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Human B5; Standard; Human B5; PE Detection Reagent; Human B5; Capture Bead B5
This item is not returnable. View return policy

Description

Description

The BD CBA Human IL-17A Flex Set is a bead-based immunoassay capable of measuring human interleukin-13 (IL-13) in serum, plasma, and cell culture supernatant samples

  • Reactivity: QC Tested: Human
  • Bead Position: B5
  • Limit of detection: 0.3pg/mL

With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), a flow cytometer, and the FCAP Array™ software

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-17A
Human B5, standard, Human B5, PE detection reagent, Human B5, capture bead B5
4°C
Human IL-17A flex set (bead B5)
Human
100 Tests
Flow Cytometry
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Cytometric Bead Array (CBA) Human IL-17A Flex Set

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research use only