missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IP-10; Flex Set, Bead B5
Bead-based immunoassay capable of measuring human interferon-Glc gamma-induced protein-10 (IP-10), also known as CXCL10, in serum, plasma, and cell culture supernatant samples
Brand: BD 558280
Includes: Human IP-10; Standard; Human IP-10; PE Detection Reagent; Human IP-10; Capture Bead B5
Description
Reactivity
- QC Testing: Human
Flow cytometry
Specifications
IP-10 | |
Human IP-10, standard, Human IP-10, PE detection reagent, Human IP-10, capture bead B5 | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Human | |
100 Tests | |
4°C | |
Human IP-10 flex set (bead B5) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Human IP-10 Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT mixture
Research Use Only