missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human TGF-β1; Flex Set, Bead B6
Human reactivity was determined by testing samples with the BD CBA Human TGF-β1 Flex Set
Brand: BD 560429
Includes: Human TGF-β1; Standard; Human TGF-β1; PE Detection Reagent; Human TGF-β1; Capture Bead B6
Description
- Capable of measuring soluble human TGF-β1 in serum and cell culture supernatant samples
Specifications
TGF-beta1 | |
Human TGF-beta1, standard, Human TGF-beta1, PE detection reagent, Human TGF-beta1, capture bead B6 | |
4°C | |
Human TGF-beta1 single plex flex set |
Human | |
100 Tests | |
Flow Cytometry |