Learn More
BD Cytometric Bead Array (CBA) Mouse IL-3; Flex Set, Bead A8
Immunoassay for measuring mouse interleukin-3 in serum and cell culture supernatant samples
Brand: BD 558346
Includes: Mouse IL-3; Standard; Mouse IL-3; PE Detection Reagent; Mouse IL-3; Capture Bead A8
Description
The BD CBA Mouse IL-3 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-3 (IL-3) in serum and cell culture supernatant samples.
- Reactivity: Mouse
- Reactivity was determined by testing samples with BD CBA Mouse IL-3 Flex set
Specifications
IL-3 | |
Mouse IL-3, standard, Mouse IL-3, PE detection reagent, Mouse IL-3, capture bead A8 | |
Serum, Cell Culture Supernatant | |
Flow Cytometry |
Mouse | |
100 Tests | |
4°C | |
Mouse IL-3 flex set (bead A8) |