missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Antimicrobic Vial A Chlortetracycline
Brand: BD 233331
Alert:
ö
Description
Specifications
SMWW | |
Antimicrobic vial A Chlortetracycline | |
Preparation of Bacto Cooke Rose Bengal Agar | |
10 mL |
2°C to 8°C | |
Powder | |
Vial |