missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Culture Media Additives: Egg Yolk Enrichment 50%
To enrich media for cultivating and isolating large variety of microorganisms and tissue cells
Brand: BD 215329
This item is not returnable.
View return policy
Description
Egg yolk enrichment 50%
- For use in SFP Agar Base and McClung Toabe Agar Base for isolation of Clostridium species based on their lecithinase activity
Specifications
50% | |
2°C to 8°C | |
Egg Yolk Enrichment | |
For use in Difco™ SFP agar base, Difco™ McClung Toabe Agar base for isolation and identification of clostridium species and for enrichment of media |