missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD14
CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. BD Horizon™ BV480 Mouse Anti-Human CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells.
326.00€ - 645.00€
Specifications
Product Type | BV480 Mouse Anti-Human CD14 |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15834829
|
BD
566190 |
25 Tests |
326.00€
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
15864779
|
BD
566141 |
100 Tests |
645.00€
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD14 | |
QC Testing: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |