missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD16
Expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. BD Horizon™ BV480 Mouse Anti-Human CD16 clone 3G8 also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomologus macaque monkeys.
Brand: BD 566108
This item is not returnable.
View return policy
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD16 | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |
100 Tests | |
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |