missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD19
CD19 is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. BD Horizon™ BV480 Mouse Anti-Human CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.
326.00€ - 785.00€
Specifications
Product Type | BV480 Mouse Anti-Human CD19 |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15894799
|
BD
566164 |
25 Tests |
326.00€
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
15814749
|
BD
566103 |
100 Tests |
785.00€
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD19 | |
QC Testing: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |