missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD3
CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. BD Horizon™ BV480 Mouse Anti-Human CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε
Brand: BD 566105
This item is not returnable.
View return policy
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD3 | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |
100 Tests | |
QC Testing: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |