missing translation for 'onlineSavingsMsg'
Learn More

BD Human IL-1β ELISA Kit II

For the quantitative determination of Human IL-1Β in serum, plasma and cell culture supernatant

Brand:  BD 557966


Product Code. 15859138

  • 1360.00€ / Each
Estimated Shipment: 18-11-2024
to see stock.
Explore more special offers
Add to basket

Includes:

Antibody Coated Wells, Detection Antibody, Standards, Enzyme Concentrate (250x), Enzyme Diluent, ELISA Dilution reagent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers

This item is not returnable. View return policy

Description

Description

  • Standard Range: 1.95-125pg/mL
  • Potent immunomodulator
  • Formulated for improved sensitivity

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-1beta
Stop solution, Human IL-beta standard, TMB one-step Substrate reagent, Enzyme diluent, Human IL-1beta Antibody coated plate, ELISA dilution reagent, detection Antibody Biotin Anti-Human IL-1beta, Enzyme concentrate Streptavidin HRP, wash concentrate (20X)
Serum, Plasma, Cell Culture Supernatant
Quantitative determination of Human IL-1beta, ELISA
Human
2 Plates
2°C to 8°C
Human IL-1beta ELISA kit II
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Human IL-1beta ELISA Kit II

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : phosphoric acid, CMIT/MIT

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only