missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse IFN-γ ELISA Set

For immunoassay protocols

Brand:  BD 555138

911.95 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15858008

  • 915.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • Capture Antibody: Anti-mouse IFN-Γ monoclonal antibody
  • Detection Antibody: Biotinylated anti-mouse IFN-Γ monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant mouse IFN-Γ, lyophilized
  • Instruction / Analysis Certificate
This item is not returnable. View return policy

Description

Description

The OptEIA set for mouse interferon-Γ (IFN-Γ) contains the components necessary to develop enzyme-linked immunosorbent assays (ELISA).

  • Reactivity: Mouse
  • For natural or recombinant mouse IFN-Γ in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IFN-gamma ELISA set
To develop ELISA for natural or recombinant Mouse IFN-g in Serum, Plasma and cell culture supernatants
20 Plates
20 96-well Plates
anti-Mouse IFN-gamma monoclonal (capture) antibody, Biotinylated anti-Mouse IFN-gamma monoclona (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IFN-g (standard), Streptavidin-HRP Conjugate
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Mouse IFN-γ ELISA Set > Reagents for 20 plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only