Learn More
BD Mouse Naive/Memory T Cell Panel
Contains fluorescent antibodies specific for CD44, CD62L, CD4 and CD3 cell surface antigens
Brand:  BD 561609
680.28 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15831169
  • 685.00€ / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Alexa Fluor™ 647 Mouse Anti-Human CD197 (CCR7), APC-H7 Mouse Anti-Human CD3, PerCP-Cy™5.5 Mouse Anti-Human CD4, FITC Mouse Anti-Human CD45RA
Description
Description
  • Designed to standardize multicolor staining and flow cytometric characterization
  • Contains antibodies optimized at 5μl per test
  • Formats and concentration optimized for each specificity to work as a combined panel
  • Compatible with dual-laser flow cytometers such as the BD FACSCalibur™
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse
50 Tests