Learn More
BD Pharmingen™ MitoStatus TMRE
In cells undergoing apoptosis, oxidative stress, necrosis, and other cellular processes, the mitochondrial membrane potential (ΔΨm) can become depolarized.
Brand:  BD 564696
195.67 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15873819
  • 196.00€ / Each
Estimated Shipment: 19-03-2025
Description
In cells undergoing apoptosis, pro-apoptotic Bcl-2 family proteins cause mitochondrial outer membrane permeabilization (MOMP), resulting in the release of cytochrome C and the subsequent activation of caspase-9 and the apoptotic cascade. MOMP often correlates with the loss of inner mitochondrial membrane potential (ΔΨm), which can be detected using ΔΨm-sensitive dyes. These cationic, lipophilic dyes accumulate within the mitochondria of healthy cells, but not within mitochondria that have lost ΔΨm due to induction of apoptosis or treatment with a mitochondrial uncoupler.
BD Pharmingen™ MitoStatus TMRE (Tetramethylrhodamine ethyl ester) is a fluorescent dye that is readily sequestered by active mitochondria, allowing for flow cytometric or imaging analysis to assess for apoptosis or mitochondrial depolarization. Non-apoptotic or cells with polarized mitochondria will fluoresce red while apoptotic or cells with depolarizard mitochondria will have diminished levels of red fluorescence. MitoStatus TMRE (Tetramethylrhodamine ethyl ester) has an excitation maximum of 549nm, but is well-excited by both the blue (e.g. 488 nM) and yellow-green (e.g. 561nM) lasers. It has an emission maximum of 574nm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mg
Lyophilized powder
Tested in Development: Human, Mouse
Upon arrival, store the dye powder desiccated and protected from light at ≤ -20°C until use. The dye powder is stable for at least 12 months if stored as indicated. After reconstitution with DMSO, store the stock solution at ≤ -20°C in small aliquots. The stock solution is stable for at least 6 months if stored as indicated.
Documents
One moment while we fetch your results.
Product Title
BD Pharmingen™ MitoStatus TMRE > 25mg