Learn More
BD Prepared Bottled Media: Reinforced Clostridial
For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples
Brand: BD 215192
This item is not returnable.
View return policy
Description
Ready for immediate use
Specifications
Amber | |
Liquid | |
Reinforced Clostridial Medium | |
Detection of microorganisms from pharmaceutical, food, environmental or Water testing samples | |
10/Pk. |
2 to 25°C | |
United States | |
100 mL | |
Bottle |