Learn More
BD Vacutainer BD Vacutainer™ EST Tube
Contains no additives and is suitable as a secondary tube for anti-coagulated blood samples.
Brand: BD Vacutainer 364917
This item is not returnable.
View return policy
Description
- Evacuated secondary tube (EST)
- No additives
- Suitable as a secondary tube for anti-coagulated blood samples, for example for taking plasma samples from blood bags
- Can also be used as a discard tube
Specifications
No Additive | |
11 mL | |
Secondary tube for anti-coagulated blood samples. |
PET | |
16 x 100 mm | |
Paper |