Description
- Non woven
- With round elastics
- White
- Polypropylene
- Latex-free
Specifications
Specifications
Type | General Purpose Beard Cover |
Material | Spunbond Polypropylene |
Color | White |
Disposable | Single-use |
Latex Free | Yes |
Packaging Quantity | 1000/Pk. |
Weight (Metric) | 10 gsm |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
Mutexil™ Beard Covers >