Learn More
behr Labor-Technik™ behropur™ Water Deionizer Made Of Nylon
Made from Nylon. Convenient and safe mixed-bed ion exchanger for small to medium-sized quantities of high purity water. To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
Brand:  behr Labor-Technik™ B00223475
 View more versions of this product
Product Code. 10544862
  • 1510.00€ / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
    • Can be connected to the raw water network directly and without a pressure reducer
      • Resistant to alternating pressure
        • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
          • Also available with shutoff when the limited value is reached and limit control
            • Pressure resistant to 8 bar
Specifications
Specifications
B10DNZ Pressure resistant mixed-bed unit made of nylon, permanent pressure resistant to 8 bar, spare cartridge
300 L/h
B10DNZ
22 cm
60 cm
1200 L
Nylon
Demineralizers
22 cm