Learn More
behr Labor-Technik™ behrotest™ Extraction Device for Twisselmann Hot Extraction System
Extractors for multi-samples.
7430.00€
Specifications
Product Type | Extraction Device |
---|---|
For Use With (Equipment) | Twisselmann Hot Extraction System |
Includes | Brackets |
For Use With (Application) | Extraction of Solvents |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10245152
|
behr Labor-Technik™
B00218424 |
6 x 100 mL |
7430.00€
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- Extraction according to Twisselmann method
- Continuous hot extraction
- Improves solubility and shortens extraction time
- Energy individually adjustable for every sample position
- Cooling water distribution strip ensures uniform cooling at sample positions
- Practical condenser ledge to safely deposit condenser between hot extractions
- Brackets for safely holding intermediate extraction pieces after removing sample container
Specifications
Extraction Device | |
Twisselmann Hot Extraction System | |
Brackets | |
Extraction of Solvents |