Learn More
Astell Scientific™ Benchtop Autoclaves
Totally self-contained autoclaves with heaters in chamber as standard, requiring no plumbing and only a single phase power supply.
Brand:  Astell Scientific™ AMB430
 View more versions of this product
Product Code. 12745375
  • 12360.00€ / Each
Estimated Shipment: 25-02-2025
Description
  • Classic version requires manual water-fill and during the cycle condensate leaves the vessel via a silicone tube that can be connected to a heat-resistant bottle
  • Media holdwarm and delayed start facility
  • A 5.7 in. colour touchscreen with integrated USB data port
  • Electropolished stainless steel chamber
  • Simple pre-post vacuum option available
  • Adjustable time and temperature
Specifications
Specifications
13 A
43 L
600 mm
Benchtop Autoclave
230 V
465 mm
346 mm
AMB
50/60 Hz
520 mm