missing translation for 'onlineSavingsMsg'
Learn More

Benzamil, Tocris Bioscience™

Product Code. 18776067
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NCX inhibitor, Deg/ENaC channel blocker; amiloride (Cat. No. 0890) derivative

Na+/Ca2+ exhanger (NCX) inhibitor (IC50 ∽ 100 nM); TRPP3 channel blocker (IC50 = 1.1 μM for inhibition of Ca2+-activated TRPP3 channel activity). Also non-selective Deg/ENaC family blocker; reduces mechanosensitivity of colonic afferents. More potent derivative of amiloride (Cat. No. 0890).

Chemical Identifiers

CAS 161804-20-2
Molecular Formula C13H14ClN7O·HCl
Synonym N-(Benzylamidino)-3, 5-diamino-6-chloropyrazinecarboxamide hydrochloride

Specifications

Quantity 50 mg
Formula Weight 356.21
Percent Purity >98%
Chemical Name or Material Benzamil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.