missing translation for 'onlineSavingsMsg'
Learn More
Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals Product Code.: 11379587

Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals

Product Code. 11379587
4 x 1 L
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11379587

Brand: Thermo Scientific Alfa Aesar 041218.M1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-51-6 | C7H8O | 108.14 g/mol

Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcoholShow MoreShow Less
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15°C
Density 1.045 g/mL
Boiling Point 205°C
Flash Point 100°C (212°F)
Odor Aromatic
Refractive Index 1.54
Quantity 4 x 1 L
Beilstein 878307
Merck Index 14,1124
Solubility Information Misicible with water,benzene,methanol,chloroform,ethanol,acetone and ether.
Formula Weight 108.14
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material Benzyl alcohol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX04K955 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number041218.M1,041218.K2,041218.K7,041218.AK
Lot NumberX04K954 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041218.M1,041218.K2,041218.K7,041218.AK
Lot NumberQ19K796 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number041218.M1,041218.K2,041218.K7,041218.AK
Lot NumberQ19K795 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number041218.M1,041218.K2,041218.K7,041218.AK
Lot NumberM12K708 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number041218.M1,041218.K2,041218.K7,041218.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.