Learn More
Buerkle™ Container Connection with Outer Thread
For the easy mounting of thread in the sides of containers.
Brand: Buerkle™ 8530-0012
10.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- With outer thread
- Complete with EPDM seals
Specifications
Container Connection | |
Polypropylene | |
13.5 mm | |
Gray |
Autoclavable | |
Used for Easy Mounting of Thread in The Sides of Containers | |
1/2 in. thread size |