Learn More
Gilac™ Biobased Dough Container
Biobased Dividing Dough Container is highly resistant to changes in temperature, highly flexible and shock-resistant.
22.55€ - 24.70€
Product Code | Brand | Volume (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
17998551
|
Gilac™
G179391 |
9 L | 600 x 400 x 55 mm |
22.55€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17968551
|
Gilac™
G179491 |
15 L | 600 x 400 x 90 mm |
24.70€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
Description
- Biobased dough storage container - 600 x 400.
- Stacking with and without lid, maximum stacking height capacity.
- Innovative design for even greater convenience:
- Rounded corners and bottom for easy cleaning.
- Vertical sides without inner edge avoids marking the dough. The entire surface of the container is usable.
- Completely flat bottom means dough detaches easily, no loss, no food deposit left.
- Optimised side reinforcements allows you to customise containers with your logo.
- Rounded outside corners guarantee a damage-free working environment.
- The dough container made from biosourced material is dye- and odour-free with a white appearance.
- Biobased material is made from natural renewable resources instead of limited energy sources such as fossil fuels.
- As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety in full compliance with current regulations.
- This dough container adds to the existing range of dough containers made with conventional materials.
Specifications
Dough Container | |
Polyethylene | |
White |