Learn More
Sartorius™ Biohit™ Proline™ Fixed Volume Pipettes
Single channel fixed volume pipettes.
Brand: Sartorius™ 722050
179.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Single channel fixed volume pipettes.
Specifications
1 | |
Fixed Volume Pipette | |
2000 μL |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%
Product Content Correction
Product Title
Sartorius™ Biohit™ Proline™ Fixed Volume Pipettes > 2000μL