Learn More
Eppendorf™ Biopur™ epT.I.P.S.™ Singles
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Eppendorf™ Biopur™ epT.I.P.S.™ Singles are the perfect combination that results in reduced-force tip attachment and ejection with complete sealing.
Brand: Eppendorf™ 0030010027
33.06 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
- For use in pharmaceutical and food industry, molecular biology and cell technology
- Batch number and expiration date on each blister package
- Each production lot inspected continuously by an independent laboratory
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
0.1 to 20 μL | |
Sterile | |
Medium gray | |
epT.I.P.S.™ Singles | |
Biopur™ | |
100 tips, individually packed | |
epT.I.P.S. |
Non-filtered | |
Natural | |
Autoclavable | |
40mm | |
Singles | |
Medium Gray |