Learn More
Sartorius™ Biosart™ Monitor
Product Code. p-8007452 Shop All Sartorius Products
missing translation for 'orderingAttributeHoverText'
Color:
Gray/White
Green/Dark Green
White/Black
Packaging:
2 layers of 24
4 bags of 12
48/Pk.
missing translation for 'unitSize'
Pack of 48
Product Code. 11823292
missing translation for 'mfr': Sartorius™ 164024706K
Microbiological and analytical filtration units offering superior performance
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
TRUSTED_SUSTAINABILITY
Specifications
Color Green/Dark Green
Sterility Sterile
No. per Case 48Pack
No. per Pack 48Pack
Packaging 4 bags of 12
Quantity 48Pack
Type Monitor
compliance-icons compliance-icons
Product Identifier
  • X48 BIOSART 100 0,45µM 47MM GREEN
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose