Learn More
Birch pollen Bet V1 Antibody (BV16), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81093PCP
Description
Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA
Specifications
Birch pollen Bet V1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Plant | |
Purified |
Western Blot, ELISA | |
BV16 | |
Western Blot, ELISA | |
Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
Birch pollen Bet V1-A, a major allergen of the European white birch. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.