Learn More
Bis(4-fluorophenyl) ether, 98%, Thermo Scientific™
41.10€ - 117.00€
Chemical Identifiers
CAS | 330-93-8 |
---|---|
Molecular Formula | C12H8F2O |
Molecular Weight (g/mol) | 206.19 |
MDL Number | MFCD00013552 |
InChI Key | UUKHFGSOCZLVJO-UHFFFAOYSA-N |
Synonym | bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro |
PubChem CID | 67615 |
IUPAC Name | 1-fluoro-4-(4-fluorophenoxy)benzene |
SMILES | FC1=CC=C(OC2=CC=C(F)C=C2)C=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
330-93-8 | |
206.19 | |
UUKHFGSOCZLVJO-UHFFFAOYSA-N | |
67615 | |
FC1=CC=C(OC2=CC=C(F)C=C2)C=C1 |
C12H8F2O | |
MFCD00013552 | |
bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro | |
1-fluoro-4-(4-fluorophenoxy)benzene |
Specifications
330-93-8 | |
240°C | |
Characteristic | |
1.5375 | |
2559168 | |
UUKHFGSOCZLVJO-UHFFFAOYSA-N | |
1-fluoro-4-(4-fluorophenoxy)benzene | |
67615 | |
98% |
1.23 | |
>110°C (230°F) | |
C12H8F2O | |
MFCD00013552 | |
bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro | |
FC1=CC=C(OC2=CC=C(F)C=C2)C=C1 | |
206.19 | |
206.19 | |
Bis(4-fluorophenyl) ether |
Safety and Handling
missing translation for 'einecsNumber' : 206-358-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.