Learn More
Fisherbrand™ Black Phenolic Caps Product Code.: 16134587
Estimated Shipment: 17-09-2025
Add to basket
Fisherbrand™ Black Phenolic Caps
Product Code. 16134587 Shop All Fisherbrand Products
20-400, 576
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Product Code. 16134587
Estimated Shipment: 17-09-2025
Add to basket
Add to basket
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Color Black
Quantity 576
Closure Color Black
Neck Finish 20-400
Closure Included No
Product Type Lined Cap
Closure Liner Polyethylene Cone
Closure Material Phenolic
Product Content Correction
Product Title
Fisherbrand™ Black Phenolic Caps >