Learn More
Fisherbrand™ Black Phenolic Caps Product Code.: 16154587
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
Fisherbrand™ Black Phenolic Caps
Product Code. 16154587
22-400, 12
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Product Code. 16154587
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
Add to basket
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Color Black
Quantity 12
Closure Color Black
Neck Finish 22-400
Closure Included No
Product Type Lined Cap
Closure Liner Polyethylene Cone
Closure Material Phenolic
Product Content Correction
Product Title
Fisherbrand™ Black Phenolic Caps >