Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Brand:  BRAND™ 43308
49.54 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10663561
  • 62.85€ / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
Estimated Shipment: 30-07-2025
Add to basket
Description
Description
  • Calibrated density bottle made with Borosilicate glass
  • DIN ISO 3507, type Gay-Lussac
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • The volume in cmÂł is specified to a precision of 3 decimal places
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2Pack
10 mL
Borosilicate Glass
Calibrated Density Bottle
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles > Capacity: 10mL