Learn More
Fisherbrand™ Blue Micro PTFE Stir Bar
Fisherbrand™ Blue Micro PTFE Stir Bar is an inert and leak-free stirrer for a wide variety of applications where identification is important.
Brand: Fisherbrand™ 001.803-B
Description
- Micro stirrer bars
- For use where identification is of prime importance
- PTFE coatings are not as inert as pure PTFE
- Individually polished to stay cleaner for longer
Specifications
Autoclavable | |
Blue | |
Containers & Vessels / Stirrer Hot Plates | |
3 mm | |
10/Pk. | |
PTFE Stirrer Bars Micro Coloured |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
PTFE | |
3 mm | |
Micro |