Learn More
Brady™ BMP71 BMP61 M611 Continuous Matte White Ultra Aggressive Polyester General ID Labels
Ultra Aggressive Polyester (B-489A) with permanent adhesive is ideal for labeling products and general ID in both commercial and industrial applications.
97.06€ - 158.57€
Specifications
Adhesive Type | Permanent Rubber Based |
---|---|
Color | White |
Finish | Matte |
Material | Polyester (B-489A) |
Print Technology | Thermal Transfer |
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
11512585
|
Brady™
011246 |
15.24 m x 6.35 mm | |||||||
11522575
|
Brady™
011236 |
15.24 m x 12.7 mm | |||||||
11562555
|
Brady™
011220 |
15.24 m x 33.02 mm | |||||||
11562575
|
Brady™
011241 |
15.24 m x 25.4 mm | |||||||
11582575
|
Brady™
011243 |
15.24 m x 48.26 mm | |||||||
Description
- Labels for the BMP61 and M611 Thermal Transfer Portable Printers
- Material is UL & CSA approved with its respective ribbon
- See the Recommended Ribbon Series for the appropriate ribbon to use
- Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Permanent Rubber Based | |
Matte | |
Thermal Transfer |
White | |
Polyester (B-489A) | |
BMP61, BMP71 (with Media Adapter), M611 |