Learn More
Thermo Scientific Chemicals Boiling stone
For preparative work
Brand:  Thermo Scientific Chemicals 386210500
22.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10340534
  • 22.51€ / 50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Boiling Stone
50 g
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Boiling stones
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461397 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number386212500,386210500
Lot NumberA0450491 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number386212500,386210500
Lot NumberA0400285 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number386212500,386210500
Lot NumberA0403698 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number386212500,386210500
Lot NumberA0407178 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number386212500,386210500
Product Title
Thermo Scientific Chemicals Boiling stone > 50g; Plastic bottle