Learn More
Bohlender™ BOLA™ Screw Caps Product Code.: 10379281
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
Bohlender™ BOLA™ Screw Caps
Product Code. 10379281
PBTP
Click to view available options
Material:
PBTP
Product Code. 10379281
Brand: Bohlender™ H98701
Estimated Shipment: 04-06-2025
Close bottles and vessels with GL 14 threads. Bohlender™ BOLA™ Screw Caps feature glass-fiber reinforced PBTP construction with integrated PTFE/silicone gasket.
  • Screw cap with handy knurl
  • Integrated PTFE/silicone gasket
  • After assembly, product is only exposed to PTFE
Specifications
Material PBTP
Temperature Range (Metric) -45 to 180 deg.C
For Use With (Equipment) GL thread 14
Product Content Correction
Product Title
Bohlender™ BOLA™ Screw Caps >