Learn More
Berkshire International™ BCR™ Bond Paper
BIOPHARMA
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
Brand:  Berkshire International™ B85A4PK25010
14.20 EUR valid until 2025-06-21
Use promo code "24097" to get your promotional price.
 View more versions of this product
Product Code. 12817143
  • 28.05€ / Pack of 250
Outlet Corner
Use promo code 24097 at checkout
View Available Promotions
In Stock
Edge
Add to basket
Description
Description
  • Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Specifications
Pink
Sheets
BCR Bond 850
A4
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0831642 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog NumberB85A4PK25010
Lot Number0000840610 Certificate TypeCertificate of Analysis Date12/10/2022 Catalog NumberB85.A4.PK.250/10
Lot Number0000840610 Certificate TypeDeclaration of Conformity Date12/10/2022 Catalog NumberB85.A4.PK.250/10
Lot Number0832301 Certificate TypeCertificate of Compliance Date13/09/2021 Catalog NumberB85.A4.PK.250/10
Promotions
Promotions
  • Multiple Manufacturers
    Use code 24097 to receive this promotion.
    Promo Code: 24097
    Expires2025-06-16
    Days
    17
    Hours
    20
    Minutes
    44
    Seconds
    57
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 19/05/2025 and 16/06/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
Product Content Correction
Product Title
Berkshire International™ BCR™ Bond Paper > Page Format: A4, Pink, Bond 850