Learn More
Boron Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™ Product Code.: 10619852
Boron Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™
Product Code. 10619852
100 mL
Click to view available options
Quantity:
100 mL
500 mL
Product Code. 10619852
Brand: Fisher Chemical J/8006/05
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
MDL Number 134034
Specifications
Packaging HDPE plastic bottle
Quantity 100 mL
Physical Form Liquid
Color Colorless
Product Content Correction
Product Title
Boron Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™ >