Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04006158
 View more versions of this product
Product Code. 11871593
  • 130.00€ / Pack of 2
Estimated Shipment: 23-05-2025
Description
  • With scale (approx. volume/mL)
  • With PTFE plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
500 mL
Yes
2
Borosilicate Glass
10mL
Polyethylene
Product Title
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale > Capacity: 500mL