Learn More
IKA™ Borosilicate Glass Evaporation Cylinder
901.00€ - 1031.00€
Specifications
Material | Borosilicate Glass |
---|---|
Type | Evaporating Cylinder |
Quantity | 1 Pack |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11885912
|
IKA™
0003738800 |
500 mL |
901.00€
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
11895912
|
IKA™
0003738900 |
1500 mL |
1031.00€
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Specifications
Borosilicate Glass | |
Evaporating Cylinder | |
1 Pack |