Learn More
Cytiva Whatman™ Borosilicate Glass Frits
Intended for use with GV-Series Vacuum Filtration Systems, Whatman™ Borosilicate Glass Frits are recommended for filtration (aqueous and organic solvents) and biological analysis.
Brand: Cytiva 10442003
69.97 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Material: Borosilicate Glass
- For Use With (Application): Filtration (aqueous and organic solvents) and biological analysis
Specifications
Reusable | |
Borosilicate Glass | |
Vacuum Filtration | |
1 Ea. |
Whatman GV050 Series Glass Vacuum Filtration System | |
Non-sterile | |
GV050/0/03 | |
Glass Frit Filter |
Product Content Correction
Product Title
Cytiva Whatman™ Borosilicate Glass Frits > Borosilicate Glass Frits; For Use With: GV050 Series Glass Vacuum Filtration System
For internal research use only. Products are not intended for diagnostic use or resale.