Learn More
Rogo Sampaic™ Glass Measuring Cup
Brand:  Rogo Sampaic™ CBD007
 View more versions of this product
Product Code. 11764136
  • 66.00€ / Each
Estimated Shipment: 04-08-2025
Specifications
Thick Glass
500 mL
190 mm
Product Title
Rogo Sampaic™ Glass Measuring Cup > Capacity: 500mL