Learn More
Soda Lime Glass Petri Dishes
High-quality petri dish, with lid.
Brand: Fisherbrand 2290/60X12
This item is not returnable.
View return policy
Description
- Constructed of soda lime glass.
- Includes lid
Specifications
Closure Included | |
Round | |
12 mm |
Soda Lime Glass | |
Petri Dish | |
60 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Soda Lime Glass Petri Dishes > Dimensions: 60 dia. x 12mmH