Learn More
Scientific Glass Labs™ Borosilicate Glass Powder Funnel
Manufactured from borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Powder Funnel is less likely to clog or cake when funneling powdery substances.
5.25€ - 8.80€
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | ||||
12993591
|
Scientific Glass Labs™
FPH-240-070U |
70mm | 16mm | 25mm |
5.25€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||
12903601
|
Scientific Glass Labs™
FPH-240-090X |
100mm | 22mm | 25mm |
8.80€
Each |
In Stock
Log in to see stock. |
||||
12983591
|
Scientific Glass Labs™
FPH-240-050D |
50mm | 13mm | 20mm |
5.25€
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
- Manufactured from borosilicate glass.
- Short stem funnel
- Less likely to clog or cake when funneling powdery substances
Specifications
Borosilicate Glass |
Product Title
Scientific Glass Labs™ Borosilicate Glass Powder Funnel