Learn More
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap
Short-style shell vials are ideal for storing dry products
Brand:  Kimble Chase™ 60931-1
 View more versions of this product
Product Code. 11823193
  • 33.10€ / Pack of 144
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
In Stock
Edge
Add to basket
Description
Description
  • Plain top design
  • Lab pack quantities
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mL
1440
144
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap > Capacity: 4mL