Learn More
Witeg™ Borosilicate Glass Tall Form Beakers
Witeg™ Borosilicate Glass Tall Form Beakers conveniently include graduations and a pouring spout.
Brand:  Witeg™ 5 502 250 S
 View more versions of this product
Product Code. 11867850
  • 25.45€ / Pack of 10
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durable borosilicate glass
  • Graduated
  • Tall form with spout
  • Some models include handles
Specifications
Specifications
61 mm
Borosilicate Glass (3.3)
Spout
Beaker
ISO 3819, DIN 12331
250 mL
117 mm
10 Pk.
Yes
Tall Form
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Witeg™ Borosilicate Glass Tall Form Beakers > Capacity: 250mL; Diameter: 61mm