Learn More
Emma Safety Footwear Boston Safety Shoes
The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable. The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
Brand: Emma Safety Footwear MM40064739
This item is not returnable.
View return policy
Description
- S1P-Safety shoe with sole plate for dry work
- Knitted Textile construction
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD Certified
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Antibacterial | |
39 |
Laces | |
Boston | |
Textile | |
Composite |
Product Content Correction
Product Title
Emma Safety Footwear Boston Safety Shoes > Size: 39