Learn More
Mutexil™ Charlotte Disposable Round Caps Product Code.: 11725443
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
Mutexil™ Charlotte Disposable Round Caps
Product Code. 11725443
53 cm, Bouffant, 500/Pk.
Click to view available options
Diameter (Metric):
24 cm
48 cm
53 cm
Product Type:
Bouffant
Quantity:
1000/Pk.
500/Pk.
Product Code. 11725443
Brand: Mutexil™ 3322090
Estimated Shipment: 20-06-2025
Charlotte caps are ideal in clinic, hospital, restaurant, and food processing settings. Eco-friendly, disposable caps are elastic and are constructed of non-woven polypropylene.
  • Round
  • Polypropylene
  • Non-woven
  • White
Specifications
Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Design Type Round
Quantity 500/Pk.
Diameter (Metric) 53 cm
Product Content Correction
Product Title
Mutexil™ Charlotte Disposable Round Caps >