Learn More
MP Biomedicals™ Casein Hydrolysate Enzymatic, Powder
Casein hydrolysate enzymatic is recommended for use as Animal diet
Brand: MP Biomedicals™ 0210129005
Description
Applications:
Casein hydrolysate enzymatic is recommended for use as Animal diet.
Product Description:
Casein hydrolysate enzymatic is a pancreatic hydrolysate of casein. This hydrolysate contains, in the form of mixed amino acids and peptides, all amino acids originally present in casein. It is a refined product providing high quality soluble source of amino acids, polypeptides and peptides.

Specifications
Animal Diet | |
Animal diet | |
Room Temperature |
Powder | |
5 lb. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S6794 | Certificate of Analysis | 17/03/2022 | 0210129005 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.