missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Bovine Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of Toxoplasmosis (TOXO) Antibody (IgG).
Brand: Novus Biologicals™ NBP2-60170-1KIT
This item is not returnable.
View return policy
Description
The Bovine Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric) measures bovine Toxoplasmosis (TOXO) Antibody (IgG) in serum.Specifications
ELISA Kit (Colorimetric) | |
Bovine | |
1 Kit | |
Cell Biology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
Serum | |
ELISA | |
RUO | |
50 to 100 μL | |
Toxoplasmosis (TOXO) Antibody (IgG) |
For Research Use Only.